Assay Details
Gene Symbol: TSPAN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046987.1 | XP_017902476.1 | 2 - 3 | 432 | 55 | |
GenBank mRNA | JT688326.1 | - | 7 - 8 | 1999 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
