Assay Details
| Gene Symbol: NUDT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683475.3 | XP_005683532.1 | 6 - 7 | 1500 | 60 | |
| XM_018049991.1 | XP_017905480.1 | 6 - 7 | 1567 | 60 | ||
| XM_018049992.1 | XP_017905481.1 | 6 - 7 | 1406 | 60 | ||
| XM_018049993.1 | XP_017905482.1 | 7 - 8 | 1546 | 60 | ||
| GenBank mRNA | JO408893.2 | - | 1 - 2 | 348 | 60 | |
More Information
Set Membership: |
|

NUDT12
Genomic Map