Assay Details
Gene Symbol: ABHD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695792.3 | XP_005695849.1 | 4 - 5 | 441 | 56 | |
XM_018038381.1 | XP_017893870.1 | 4 - 5 | 523 | 56 | ||
XM_018038382.1 | XP_017893871.1 | 4 - 5 | 518 | 56 | ||
XM_018038383.1 | XP_017893872.1 | 4 - 5 | 399 | 56 | ||
XM_018038384.1 | XP_017893873.1 | 4 - 5 | 557 | 56 | ||
XM_018038385.1 | XP_017893874.1 | 3 - 4 | 458 | 56 | ||
GenBank mRNA | JO640054.2 | - | 3 - 4 | 457 | 56 | |
JO640056.2 | - | 4 - 5 | 522 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
