Assay Details
Gene Symbol: MRPS31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687480.3 | XP_005687537.2 | 3 - 4 | 751 | 75 | |
GenBank mRNA | JO437335.2 | - | 4 - 5 | 627 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
