Assay Details
Gene Symbol: RAB30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699478.3 | XP_005699535.1 | 2 - 3 | 337 | 61 | |
GenBank mRNA | JI861847.1 | - | 1 - 2 | 96 | 61 | |
JT644639.1 | - | 3 - 4 | 292 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
