Assay Details
Gene Symbol: KCMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055388.1 | XP_017910877.1 | 6 - 7 | 1052 | 80 | |
GenBank mRNA | KA324479.1 | - | 1 - 2 | 1345 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
