Assay Details
Gene Symbol: USB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062059.1 | XP_017917548.1 | 5 - 6 | 710 | 58 | |
XM_018062060.1 | XP_017917549.1 | 5 - 6 | 710 | 58 | ||
XM_018062061.1 | XP_017917550.1 | 5 - 6 | 710 | 58 | ||
XM_018062062.1 | XP_017917551.1 | 5 - 6 | 1025 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
