Assay Details
| Gene Symbol: PCYOX1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050178.1 | XP_017905667.1 | 3 - 4 | 522 | 76 | |
| GenBank mRNA | JO608948.2 | - | 3 - 4 | 1248 | 76 | |
More Information
Set Membership: |
|

PCYOX1L
Genomic Map