Assay Details
Gene Symbol: OSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684598.3 | XP_005684655.2 | 1 - 2 | 499 | 70 | |
XM_018053099.1 | XP_017908588.1 | 1 - 2 | 482 | 70 | ||
GenBank mRNA | JT704227.1 | - | 1 - 2 | 466 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
