Assay Details
| Gene Symbol: ARL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055650.1 | XP_017911139.1 | 6 - 7 | 931 | 88 | |
| XM_018055720.1 | XP_017911209.1 | 6 - 7 | 931 | 88 | ||
| GenBank mRNA | JO440795.2 | - | 6 - 7 | 88 | ||
| JT659270.1 | - | 6 - 7 | 573 | 88 | ||
| JT716497.1 | - | 6 - 7 | 88 | |||
| KA308494.1 | - | 2 - 3 | 131 | 88 | ||
More Information
Set Membership: |
|

ARL6
Genomic Map