Assay Details
| Gene Symbol: NR4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063204.1 | XP_017918693.1 | 4 - 5 | 1426 | 60 | |
| XM_018063207.1 | XP_017918696.1 | 3 - 4 | 2081 | 60 | ||
| XM_018063212.1 | XP_017918701.1 | 4 - 5 | 1425 | 60 | ||
| XM_018063219.1 | XP_017918708.1 | 4 - 5 | 1426 | 60 | ||
| XM_018063224.1 | XP_017918713.1 | 4 - 5 | 1184 | 60 | ||
More Information
Set Membership: |
|

NR4A2
Genomic Map