Assay Details
| Gene Symbol: ACHE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040581.1 | XP_017896070.1 | 2 - 3 | 2332 | 94 | |
| XM_018040582.1 | XP_017896071.1 | 2 - 3 | 2332 | 94 | ||
| XM_018040583.1 | XP_017896072.1 | 2 - 3 | 2329 | 94 | ||
| XM_018040584.1 | XP_017896073.1 | 2 - 3 | 1181 | 94 | ||
| XM_018040585.1 | XP_017896074.1 | 2 - 3 | 1201 | 94 | ||
| XM_018040586.1 | XP_017896075.1 | 2 - 3 | 1252 | 94 | ||
More Information
Set Membership: |
|

ACHE
Genomic Map