Assay Details
Gene Symbol: MAGOHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680835.2 | XP_005680892.1 | 3 - 4 | 340 | 121 | |
XM_013964209.2 | XP_013819663.1 | 3 - 4 | 277 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
