Assay Details
| Gene Symbol: ARHGDIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040544.1 | XP_017896033.1 | 5 - 6 | 709 | 77 | |
| XM_018040545.1 | XP_017896034.1 | 5 - 6 | 564 | 77 | ||
| GenBank mRNA | JO643834.2 | - | 5 - 6 | 871 | 77 | |
More Information
Set Membership: |
|

ARHGDIG
Genomic Map