Assay Details
Gene Symbol: LOC102177399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041562.1 | XP_017897051.1 | 6 - 7 | 607 | 160 | |
XM_018041563.1 | XP_017897052.1 | 6 - 7 | 524 | 160 | ||
XM_018041564.1 | XP_017897053.1 | 5 - 6 | 537 | 160 | ||
GenBank mRNA | JO609927.2 | - | 6 - 7 | 547 | 160 | |
JO616787.2 | - | 9 - 10 | 676 | 160 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
