Assay Details
Gene Symbol: MRPS30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065760.1 | XP_017921249.1 | 4 - 5 | 1063 | 87 | |
GenBank mRNA | JO434749.2 | - | 4 - 5 | 1037 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
