Assay Details
| Gene Symbol: TMEM120A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040402.1 | XP_017895891.1 | 7 - 8 | 748 | 58 | |
| XM_018040403.1 | XP_017895892.1 | 5 - 6 | 603 | 58 | ||
More Information
Set Membership: |
|

TMEM120A
Genomic Map