Assay Details
Gene Symbol: WDR83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051280.1 | XP_017906769.1 | 4 - 5 | 455 | 62 | |
GenBank mRNA | JO592762.2 | - | 4 - 5 | 364 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
