Assay Details
Gene Symbol: RAB3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044093.1 | XP_017899582.1 | 4 - 5 | 592 | 77 | |
GenBank mRNA | JT646517.1 | - | 5 - 6 | 550 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
