Assay Details
| Gene Symbol: YWHAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057913.1 | XP_017913402.1 | 4 - 5 | 776 | 63 | |
| GenBank mRNA | JO496783.2 | - | 4 - 5 | 775 | 63 | |
More Information
Set Membership: |
|

YWHAB
Genomic Map