Assay Details
Gene Symbol: LOC102180115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043681.1 | XP_017899170.1 | 666 | 82 | ||
GenBank mRNA | JO420746.2 | - | 525 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
