Assay Details
| Gene Symbol: LTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039176.1 | XP_017894665.1 | 2 - 3 | 1060 | 87 | |
| XM_018039177.1 | XP_017894666.1 | 3 - 4 | 672 | 87 | ||
| GenBank mRNA | JO605634.2 | - | 3 - 4 | 1322 | 87 | |
More Information
Set Membership: |
|

LTA
Genomic Map