Assay Details
Gene Symbol: CLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059047.1 | XP_017914536.1 | 2 - 3 | 1137 | 90 | |
XM_018059048.1 | XP_017914537.1 | 2 - 3 | 1059 | 90 | ||
XM_018059049.1 | XP_017914538.1 | 2 - 3 | 815 | 90 | ||
XM_018059050.1 | XP_017914539.1 | 2 - 3 | 1222 | 90 | ||
XM_018059051.1 | XP_017914540.1 | 2 - 3 | 1202 | 90 | ||
XM_018059052.1 | XP_017914541.1 | 2 - 3 | 662 | 90 | ||
XM_018059053.1 | XP_017914542.1 | 2 - 3 | 692 | 90 | ||
XM_018059054.1 | XP_017914543.1 | 2 - 3 | 715 | 90 | ||
GenBank mRNA | JT762540.1 | - | 1 - 2 | 1530 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
