Assay Details
Gene Symbol: FBXL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049744.1 | XP_017905233.1 | 8 - 9 | 1239 | 73 | |
XM_018049745.1 | XP_017905234.1 | 8 - 9 | 1214 | 73 | ||
GenBank mRNA | KA346587.1 | - | 8 - 9 | 1185 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
