Assay Details
| Gene Symbol: HSD17B7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677120.3 | XP_005677177.3 | 7 - 8 | 1080 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSD17B7| Gene Symbol: HSD17B7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677120.3 | XP_005677177.3 | 7 - 8 | 1080 | 62 | |
Set Membership: |
|