Assay Details
| Gene Symbol: LPGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060752.1 | XP_017916241.1 | 8 - 9 | 2992 | 83 | |
| XM_018060753.1 | XP_017916242.1 | 7 - 8 | 1367 | 83 | ||
| XM_018060754.1 | XP_017916243.1 | 8 - 9 | 2724 | 83 | ||
| XM_018060755.1 | XP_017916244.1 | 8 - 9 | 1545 | 83 | ||
| XM_018060756.1 | XP_017916245.1 | 7 - 8 | 1015 | 83 | ||
| XM_018060757.1 | XP_017916246.1 | 7 - 8 | 1363 | 83 | ||
| XM_018060758.1 | XP_017916247.1 | 8 - 9 | 1355 | 83 | ||
| XM_018060759.1 | XP_017916248.1 | 7 - 8 | 1173 | 83 | ||
| XM_018060760.1 | XP_017916249.1 | 7 - 8 | 1107 | 83 | ||
| GenBank mRNA | JO420795.2 | - | 7 - 8 | 1428 | 83 | |
| JO611580.2 | - | 7 - 8 | 1316 | 83 | ||
More Information
Set Membership: |
|

LPGAT1
Genomic Map