Assay Details
| Gene Symbol: LY86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696896.3 | XP_005696953.1 | 1 - 2 | 191 | 65 | |
| GenBank mRNA | JO331196.2 | - | 1 - 2 | 193 | 65 | |
| JT669891.1 | - | 4 - 5 | 283 | 65 | ||
More Information
Set Membership: |
|

LY86
Genomic Map