Assay Details
| Gene Symbol: IGFBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285566.1 | NP_001272495.1 | 3 - 4 | 688 | 76 | |
| XM_018054247.1 | XP_017909736.1 | 3 - 4 | 1553 | 76 | ||
| XM_018054254.1 | XP_017909743.1 | 3 - 4 | 1550 | 76 | ||
| XM_018054265.1 | XP_017909754.1 | 3 - 4 | 1498 | 76 | ||
| GenBank mRNA | JF720883.1 | - | 3 - 4 | 688 | 76 | |
| JO538184.2 | - | 3 - 4 | 1268 | 76 | ||
| KC485342.1 | - | 3 - 4 | 691 | 76 | ||
More Information
Set Membership: |
|

IGFBP5
Genomic Map