Assay Details
Gene Symbol: NECAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061775.1 | XP_017917264.1 | 5 - 6 | 485 | 61 | |
XM_018061776.1 | XP_017917265.1 | 4 - 5 | 402 | 61 | ||
GenBank mRNA | JO630566.2 | - | 5 - 6 | 518 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
