Assay Details
Gene Symbol: ZNF184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696712.3 | XP_005696769.1 | 4 - 5 | 487 | 100 | |
XM_013974056.2 | XP_013829510.1 | 4 - 5 | 593 | 100 | ||
GenBank mRNA | JO456986.2 | - | 1 - 2 | 100 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
