Assay Details
| Gene Symbol: ALDOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697671.2 | XP_005697728.1 | 6 - 7 | 863 | 64 | |
| XM_005697674.3 | XP_005697731.1 | 6 - 7 | 826 | 64 | ||
| XM_013974696.1 | XP_013830150.1 | 7 - 8 | 745 | 64 | ||
| XM_018039935.1 | XP_017895424.1 | 6 - 7 | 900 | 64 | ||
| GenBank mRNA | JO579149.1 | - | 3 - 4 | 738 | 64 | |
More Information
Set Membership: |
|

ALDOA
Genomic Map