Assay Details
| Gene Symbol: TARDBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690688.3 | XP_005690745.1 | 4 - 5 | 665 | 66 | |
| XM_013970144.2 | XP_013825598.1 | 4 - 5 | 665 | 66 | ||
| GenBank mRNA | KA353517.1 | - | 1711 | 66 | ||
More Information
Set Membership: |
|

TARDBP
Genomic Map