Assay Details
Gene Symbol: TINF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685331.3 | XP_005685388.3 | 1 - 2 | 930 | 55 | |
XM_018054625.1 | XP_017910114.1 | 1 - 2 | 930 | 55 | ||
XM_018054626.1 | XP_017910115.1 | 2 - 3 | 777 | 55 | ||
GenBank mRNA | JR119421.1 | - | 612 | 55 | ||
KA311940.1 | - | 1 - 2 | 623 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
