Assay Details
| Gene Symbol: BRI3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061405.1 | XP_017916894.1 | 2 - 3 | 431 | 60 | |
| GenBank mRNA | JO430736.2 | - | 2 - 3 | 444 | 60 | |
More Information
Set Membership: |
|

BRI3BP
Genomic Map