Assay Details
Gene Symbol: MANF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038225.1 | XP_017893714.1 | 2 - 3 | 299 | 62 | |
GenBank mRNA | JO581274.1 | - | 2 - 3 | 330 | 62 | |
KA317798.1 | - | 2 - 3 | 272 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
