Assay Details
| Gene Symbol: NRSN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057668.1 | XP_017913157.1 | 2 - 3 | 240 | 63 | |
| GenBank mRNA | JO633001.2 | - | 2 - 3 | 1421 | 63 | |
More Information
Set Membership: |
|

NRSN2
Genomic Map