Assay Details
Gene Symbol: NFE2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064944.1 | XP_017920433.1 | 4 - 5 | 1455 | 66 | |
XM_018064945.1 | XP_017920434.1 | 4 - 5 | 1477 | 66 | ||
XM_018064946.1 | XP_017920435.1 | 4 - 5 | 1422 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
