Assay Details
Gene Symbol: SH2D2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046309.1 | XP_017901798.1 | 4 - 5 | 556 | 51 | |
XM_018046310.1 | XP_017901799.1 | 4 - 5 | 556 | 51 | ||
GenBank mRNA | JO603506.2 | - | 3 - 4 | 461 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
