Assay Details
Gene Symbol: WASHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048642.1 | XP_017904131.1 | 7 - 8 | 1130 | 69 | |
GenBank mRNA | JO344048.2 | - | 7 - 8 | 984 | 69 | |
JO643210.2 | - | 7 - 8 | 1087 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
