Assay Details
Gene Symbol: MRPS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677677.2 | XP_005677734.1 | 2 - 3 | 184 | 63 | |
XM_005677678.3 | XP_005677735.1 | 2 - 3 | 227 | 63 | ||
XM_018046031.1 | XP_017901520.1 | 2 - 3 | 278 | 63 | ||
GenBank mRNA | KA332493.1 | - | 2 - 3 | 229 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
