Assay Details
| Gene Symbol: MCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046042.1 | XP_017901531.1 | 2 - 3 | 1122 | 86 | |
| GenBank mRNA | KA318919.1 | - | 2 - 3 | 1175 | 86 | |
More Information
Set Membership: |
|

MCL1
Genomic Map