Assay Details
| Gene Symbol: TRAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064320.1 | XP_017919809.1 | 3 - 4 | 401 | 66 | |
| GenBank mRNA | JO585865.2 | - | 3 - 4 | 317 | 66 | |
More Information
Set Membership: |
|

TRAF4
Genomic Map