Assay Details
Gene Symbol: RAB8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287569.1 | NP_001274498.1 | 6 - 7 | 481 | 94 | |
GenBank mRNA | JO431072.2 | - | 2 - 3 | 1422 | 94 | |
JQ407044.1 | - | 6 - 7 | 481 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
