Assay Details
Gene Symbol: ZFP91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059035.1 | XP_017914524.1 | 5 - 6 | 721 | 68 | |
GenBank mRNA | JO430053.2 | - | 5 - 6 | 532 | 68 | |
JO632649.2 | - | 5 - 6 | 875 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
