Assay Details
| Gene Symbol: RPL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693883.3 | XP_005693940.1 | 2 - 3 | 188 | 59 | |
| XM_005693884.3 | XP_005693941.1 | 2 - 3 | 188 | 59 | ||
| XM_005693885.3 | XP_005693942.1 | 2 - 3 | 285 | 59 | ||
| XM_018065160.1 | XP_017920649.1 | 2 - 3 | 286 | 59 | ||
| GenBank mRNA | JO571040.2 | - | 2 - 3 | 144 | 59 | |
| KA320940.1 | - | 3 - 4 | 475 | 59 | ||
More Information
Set Membership: |
|

RPL27
Genomic Map