Assay Details
| Gene Symbol: ABL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690923.3 | XP_005690980.1 | 10 - 11 | 1670 | 63 | |
| XM_005690924.3 | XP_005690981.1 | 10 - 11 | 1670 | 63 | ||
| XM_018060654.1 | XP_017916143.1 | 10 - 11 | 1964 | 63 | ||
| XM_018060655.1 | XP_017916144.1 | 10 - 11 | 1964 | 63 | ||
| GenBank mRNA | JT696308.1 | - | 6 - 7 | 2051 | 63 | |
More Information
Set Membership: |
|

ABL2
Genomic Map