Assay Details
Gene Symbol: TGFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314142.1 | NP_001301071.1 | 1 - 2 | 352 | 60 | |
XM_018061650.1 | XP_017917139.1 | 1 - 2 | 1237 | 60 | ||
GenBank mRNA | JQ085593.1 | - | 1 - 2 | 352 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
