Assay Details
| Gene Symbol: FBXW4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698404.3 | XP_005698461.3 | 5 - 6 | 2907 | 62 | |
| XM_018041142.1 | XP_017896631.1 | 5 - 6 | 3663 | 62 | ||
| GenBank mRNA | JO618364.2 | - | 7 - 8 | 825 | 62 | |
More Information
Set Membership: |
|

FBXW4
Genomic Map