Assay Details
Gene Symbol: TMED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061365.1 | XP_017916854.1 | 4 - 5 | 620 | 89 | |
XM_018061366.1 | XP_017916855.1 | 3 - 4 | 596 | 89 | ||
GenBank mRNA | JO429437.2 | - | 3 - 4 | 584 | 89 | |
JO497430.2 | - | 3 - 4 | 699 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
