Assay Details
Gene Symbol: ZSCAN21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039895.1 | XP_017895384.1 | 2 - 3 | 658 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
